missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKX Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16651-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a serine threonine protein kinase that has similarity to the catalytic subunit of cyclic AMP dependent protein kinases. The encoded protein is developmentally regulated and may be involved in renal epithelial morphogenesis. This protein may also be involved in macrophage and granulocyte maturation. Abnormal recombination between this gene and a related pseudogene on chromosome Y is a frequent cause of sex reversal disorder in XX males and XY females. Pseudogenes of this gene are found on chromosomes X, 15 and Y.Specifications
| PRKX | |
| Polyclonal | |
| Unconjugated | |
| PRKX | |
| PKX1, PRKX, Protein kinase PKX1, protein kinase, X linked | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19108, 501563, 5613 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P51817, Q922R0 | |
| Prkx | |
| PRKX Fusion Protein Ag10083 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction