missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKRIP1, Mouse anti-Human, Clone: M2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00079706-M08.50ug
This item is not returnable.
View return policy
Description
Sequence: MASPAASSVRPPRPKKEPQTLVIPKNAAEEQKLKLERLMKNPDKAVPIPEKMSEWAPRPPPEFVRDVMGSSAGAGSGEFHVYRHLRRREYQRQDYMDAMAEKQKLYAEFQKRLEKNKIAAEEQTAKRRKKRQKLKEKKLLAKKMKLEQKKQEGPGQPKEQGSSSSAEASGTEEEEEVPSFTMGRSpecifications
| PRKRIP1 | |
| Monoclonal | |
| Unconjugated | |
| PRKR interacting protein 1 (IL11 inducible) | |
| C114/FLJ13902/FLJ40957 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin) | |
| M2 | |
| In 1x PBS, pH 7.4 | |
| BC014298 | |
| PRKRIP1 | |
| PRKRIP1 (AAH14298, 1 a.a. ∼ 184 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 79706 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction