missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKRIP1 Monoclonal Antibody (4D11-3F11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522590
This item is not returnable.
View return policy
Description
Peptide Sequence: MASPAASSVR PPRPKKEPQT LVIPKNAAEE QKLKLERLMK NPDKAVPIPE KMSEWAPRPP PEFVRDVMGS SAGAGSGEFH VYRHLRRREY QRQDYMDAMA EKQKLYAEFQ KRLEKNKIAA EEQTAKRRKK RQKLKEKKLL AKKMKLEQKK QEGPGQPKEQ GSSSSAEASG TEEEEEVPSF TMGR.
Required for pre-mRNA splicing as component of the spliceosome (PubMed:28502770, PubMed:30705154). Binds double-stranded RNA. Inhibits EIF2AK2 kinase activity. [UniProt].
Specifications
| PRKRIP1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9H875 | |
| PRKRIP1 | |
| PRKRIP1 (AAH14298, 1 a.a. ~ 184 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 4D11-3F11 | |
| Unconjugated | |
| PRKRIP1 | |
| C114; KRAB box domain containing 3; KRBOX3; likely ortholog of mouse C114 dsRNA-binding protein; PRKR interacting protein 1 (IL11 inducible); PRKR-interacting protein 1; PRKRIP1; Src41; TK5 | |
| Mouse | |
| Protein A | |
| RUO | |
| 79706 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction