missing translation for 'onlineSavingsMsg'
Learn More

PRKRA Mouse, anti-Human, Clone: 1D4C3, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen PRKRA
Clone 1D4C3
Concentration 1000 μg/mL
Applications Western Blot, ELISA
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18065475
View Documents
Proteintech
68518-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18074475
View Documents
Proteintech
68518-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The PRKRA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PRKRA. This antibody recognizes human, mouse antigen. The PRKRA antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

PRKRA
1000 μg/mL
Liquid
Human, Mouse
DYT16, HSD 14, HSD14, PACT, PKR associated protein X, PKR associating protein X, PRKRA, RAX
Primary
1D4C3
Western Blot, ELISA
Mouse
8575
IgG2b
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.