Learn More
Invitrogen™ Human PRKG2 (PKG2), GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length
Specifications
Specifications
| Accession Number | Q13237 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 5593 |
| Molecular Weight (g/mol) | 116.6 kDa |
| Name | Human PRKG2 (PKG2), GST Tag |
| Quantity | 100 μg |
| Regulatory Status | RUO - research use only |
| Gene Alias | AW212535; cGK 2; cGK2; cGKII; cGK-II; cGMP dependent protein kinase type II; cGMP-dependent protein kinase 2; cGMP-dependent protein kinase II; EC 2.7.11.12; GDPKII; PRKG2; PRKGR2; protein kinase cGMP- dependent type 2; protein kinase, cGMP- dependent, type 2; protein kinase, cGMP- dependent, type II; protein kinase, cGMP-dependent, type II; testicular tissue protein Li 99; type II cGMP-dependent protein kinase |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.