Learn More
PRKG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21646-1-AP-20UL
 View more versions of this product
Product Code. 16871255
  • £135.00 / 20µL
Description
Phosphorylates PPP1R12A.
Specifications
Specifications
PRKG1
Polyclonal
Unconjugated
PRKG1
cGK 1, cGK1, CGKI, cGKI alpha, cGKI BETA, PGK, PKG, PRKG1, PRKG1B, PRKGR1A, PRKGR1B, Protein Kinase G1 alpha
Rabbit
Antigen Affinity Chromatography
RUO
19091, 54286, 5592
-20°C
Liquid
Western Blot, Immunoprecipitation
0.51 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P0C605, Q13976
Prkg1
PRKG1 Fusion Protein Ag16333
20 μL
Primary
Human, Rat, Mouse