missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals PRKD3/nPKC nu Antibody (4G7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00023683-M01
This item is not returnable.
View return policy
Description
PRKD3/nPKC nu Monoclonal antibody specifically detects PRKD3/nPKC nu in Human samples. It is validated for Western Blot, ELISA
Specifications
| PRKD3/nPKC nu | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| EC 2.7.11, EC 2.7.11.13, EPK2PKD3, nPKC-nu, PKC-NU, PRKCNnPKC-NU, Protein kinase C nu type, protein kinase C, nu, protein kinase D3, Protein kinase EPK2, protein-serine/threonine kinase, serine/threonine-protein kinase D3 | |
| PRKD3 (AAH30706, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSANNSPPSAQKSVLPTAIPAVLPAASPCSSPKTGLSARLSNGSFSAPSLTNSRGSVHTVSFLLQIGLTRESVTIEAQELSLSAVKDLVCSIVYQKFPEC | |
| 0.1 mg | |
| Cancer, Diabetes Research, Protein Kinase, Signal Transduction, Transcription Factors and Regulators | |
| 23683 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 4G7 | |
| Western Blot 1:500 | |
| AAH30706 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction