missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA124582
This item is not returnable.
View return policy
Description
PA1-24582 detects Protein Kinase D2 in human samples.
PRKD2 (PKD2) is a member of the protein kinase D serine/threonine protein kinase family. PRKD2 is a component in the signal transduction pathway induced by phorbol esters and the CCK b gastrin receptor. It exhibits the strongest homology to the serine threonine protein kinases PKD/PKC and PKCnu, particularly in the duplex zinc finger-like cysteine-rich motif, in the pleckstrin homology domain and in the protein kinase domain. In contrast, it shows only a low degree of sequence similarity to other members of the PKC family.
Specifications
| PRKD2 | |
| Polyclonal | |
| Unconjugated | |
| PRKD2 | |
| AI325941; HSPC187; nPKC-D2; PKD2; Prkd2; protein kinase D2; RGD1308054; serine/threonine-protein kinase D2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25865 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9BZL6 | |
| PRKD2 | |
| Synthetic 18 amino acid peptide from internal region of human PRKD2. | |
| 25 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction