missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKCSH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119836
This item is not returnable.
View return policy
Description
Positive Control: Hela cell lysate, Jurkat cell lysate, A431 cell lysate, rat kidney tissue lysate, rat liver tissue lysate, human liver tissue, human kidney tissue. Subcellular Location: Endoplasmic reticulum.
SET domain-containing proteins, such as SMYD2, catalyze lysine methylation.
Specifications
| PRKCSH | |
| Polyclonal | |
| Unconjugated | |
| PRKCSH | |
| 80K-H; 80K-H protein; advanced glycation end-product receptor 2; AGE-binding receptor 2; AGE-R2; alpha glucosidase II, beta-subunit; carbohydrate processing enzyme of the endoplasmic reticulum; G19P1; GIIB; GLU2B; glucosidase 2 subunit beta; glucosidase II; glucosidase II beta-subunit; glucosidase II subunit beta; hepatocystin; PCLD; PKCSH; PLD1; PRKCSH; Protein kinase C substrate 60.1 kDa protein heavy chain; protein kinase C substrate 80K-H; protein kinase C substrate, 80 Kda protein; VASAP-60 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 300445, 5589 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P14314 | |
| PRKCSH | |
| Recombinant protein within human Glucosidase 2 subunit beta aa 30-230. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction