missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKCSH Monoclonal Antibody (A6H9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544983
This item is not returnable.
View return policy
Description
PRKCSH Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
SET domain-containing proteins, such as SMYD2, catalyze lysine methylation.
Specifications
| PRKCSH | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P14314 | |
| PRKCSH | |
| Recombinant protein within human Glucosidase 2 subunit beta aa 51-250. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| A6H9 | |
| Unconjugated | |
| PRKCSH | |
| 80K-H; 80K-H protein; advanced glycation end-product receptor 2; AGE-binding receptor 2; AGE-R2; alpha glucosidase II, beta-subunit; carbohydrate processing enzyme of the endoplasmic reticulum; G19P1; GIIB; GLU2B; glucosidase 2 subunit beta; glucosidase II; glucosidase II beta-subunit; glucosidase II subunit beta; hepatocystin; PCLD; PKCSH; PLD1; PRKCSH; Protein kinase C substrate 60.1 kDa protein heavy chain; protein kinase C substrate 80K-H; protein kinase C substrate, 80 Kda protein; VASAP-60 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5589 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction