Learn More
Invitrogen™ PRKCQ (PKC theta) Recombinant Human Protein
PRKCQ (PKC theta)
Brand: Invitrogen™ PR4143B
Code : NEW
Additional Details : Weight : 0.01000kg
Description
In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.
Specifications
NP_006248 | |
84.4 kDa | |
PRKCQ (PKC theta) | |
Human | |
His Tag | |
Ser⁄Thr Kinases: AGC Group | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
Kinase Assay | |
100 μg | |
PKC Kinase Family | |
Recombinant | |
Storage Buffer: 20mM HEPES, pH 7.5, 150mM NaCl, 0.5mM EGTA, 0.5mM EDTA, 0.05% NP-40, 5mM DTT, 20% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.