missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKCQ (PKC theta) Recombinant Human Protein
PRKCQ (PKC theta)
Brand: Invitrogen™ P2996
Code : 50
Additional Details : Weight : 3.36000kg
Description
The PKC family of serine⁄threonine kinases, including PRKCQ (PKC theta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.Specifications
NP_006248 | |
5584, 5588 | |
5μg | |
PKC Kinase Family | |
Recombinant | |
Storage Buffer: 20mM HEPES, pH 7.5, 150mM NaCl, 0.5mM EGTA, 0.5mM EDTA, 0.05% NP-40, 5mM DTT, 20% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: AGC Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
protein kinase C, theta | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
84.4 kDa | |
PRKCQ (PKC theta) | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
PRKCQ | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
For Research Use Only. Not for use in diagnostic procedures.