missing translation for 'onlineSavingsMsg'
Learn More

PRKCH (PKC eta) Recombinant Human Protein

Product Code. 10073392
Click to view available options
Quantity:
5 μg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10073392

Brand: Invitrogen P2633

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Accession Number NP_006246
For Use With (Application) Kinase Assay
Gene ID (Entrez) 23683, 5583
Molecular Weight (g/mol) 82.2 kDa
Quantity 5 μg
Gene Alias PRKCH (PKC eta)
KinaseFamily PKC Kinase Family
Species Human
Recombinant Recombinant
Protein Tag His Tag
Content And Storage Storage Buffer: 20mM HEPES, pH 7.4, 250mM NaCl, 50% glycerol, 2mM EGTA, 2mM EDTA, 0.05% Triton X-100, 5mM DTT
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Assay Entry Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: AGC Group
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Zin-LYTE™, Omnia™ Bulk Substrate
Target Entry PRKCH
Target Gene protein kinase C, eta
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.