missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKCH (PKC eta) Recombinant Human Protein
The PKC family of serine/threonine kinases, including PRKCH (PKC eta), is activated intracellularly by signal transduction pathways.
£1434.00 - £6180.00
Specifications
Accession Number | NP_006246 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 82.2 kDa |
Gene Alias | PRKCH (PKC eta) |
KinaseFamily | PKC Kinase Family |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15458383
|
Invitrogen™
PR3588A |
100μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10615203
|
Invitrogen™
PV3608 |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent.
Specifications
NP_006246 | |
82.2 kDa | |
PKC Kinase Family | |
Recombinant | |
Storage Buffer: 20mM HEPES, pH 7.4, 250mM NaCl, 50% glycerol, 2mM EGTA, 2mM EDTA, 0.05% Triton X-100, 5mM DTT Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Baculovirus | |
Recombinant, Full Length | |
Signal Transduction |
Kinase Assay | |
PRKCH (PKC eta) | |
Human | |
His Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Serine/Threonine Kinases |