Learn More
PRKCDBP Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16250-1-AP-20UL
Description
The PRKCDBP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PRKCDBP. This antibody recognizes human, mouse, rat antigen. The PRKCDBP antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
| PRKCDBP | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 112464 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
| 313 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |