missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKCDBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599979
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PRKCDBP.
JAB-1 is a 38 kDa protein which was originally described as a coactivator for cJUN. It is present in a large protein complex called the Signalosome, which has been proposed to play a signaling role in mammalian cells. This protein complex appears to be analogous to the COP9 complex in plants, which is important in light-mediated plant development. Although the specific function of JAB-1 in this complex is unclear, it has recently been implicated in targeting proteins for degradation by the proteasome.
Specifications
| PRKCDBP | |
| Polyclonal | |
| Unconjugated | |
| CAVIN3 | |
| 3110015B12Rik; 6330514M23Rik; Caveolae-associated protein 3; cavin 3; CAVIN3; cavin-3; D3T-inducible gene 2 protein; Dig2; DIG-2; Dithiolethione-inducible gene 2 protein; dithiolethione-inducible gene-2; hSRBC; MGC20400; PKC-delta binding protein; Prkcdbp; protein kinase C delta binding protein; protein kinase C delta-binding protein; protein kinase C, delta binding protein; sdr-elated gene product that binds to c-kinase; sdr-related gene product that binds to c-kinase; serum deprivation response factor-related gene product that binds to C-kinase; SRBC | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 109042, 112464, 85332 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q91VJ2, Q969G5, Q9Z1H9 | |
| CAVIN3 | |
| A synthesized peptide derived from human CAVIN3(Accession Q969G5), corresponding to amino acid residues E159-P209. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction