missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKCDBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531411
This item is not returnable.
View return policy
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (88%), Rat (90%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
JAB-1 is a 38 kDa protein which was originally described as a coactivator for cJUN. It is present in a large protein complex called the 'Signalosome', which has been proposed to play a signaling role in mammalian cells. This protein complex appears to be analogous to the COP9 complex in plants, which is important in light-mediated plant development. Although the specific function of JAB-1 in this complex is unclear, it has recently been implicated in targeting proteins for degradation by the proteasome.
Specifications
| PRKCDBP | |
| Polyclonal | |
| Unconjugated | |
| CAVIN3 | |
| 3110015B12Rik; 6330514M23Rik; Caveolae-associated protein 3; cavin 3; CAVIN3; cavin-3; D3T-inducible gene 2 protein; Dig2; DIG-2; Dithiolethione-inducible gene 2 protein; dithiolethione-inducible gene-2; hSRBC; MGC20400; PKC-delta binding protein; Prkcdbp; protein kinase C delta binding protein; protein kinase C delta-binding protein; protein kinase C, delta binding protein; sdr-elated gene product that binds to c-kinase; sdr-related gene product that binds to c-kinase; serum deprivation response factor-related gene product that binds to C-kinase; SRBC | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 112464 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q969G5 | |
| CAVIN3 | |
| Recombinant protein encompassing a sequence within the center region of human PRKCDBP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction