missing translation for 'onlineSavingsMsg'
Learn More

PRKCDBP Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18418636 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418636 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418636 Supplier Novus Biologicals Supplier No. NBP281847AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRKCDBP Polyclonal antibody specifically detects PRKCDBP in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKCDBP
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias Cavin-3, hSRBC, MGC20400, protein kinase C, delta binding protein, sdr-related gene product that binds to c-kinase, Serum deprivation response factor-related gene product that binds to C-kinase, SRBCprotein kinase C delta-binding protein
Host Species Rabbit
Immunogen Rabbit polyclonal PRKCDBP antibody was raised against an 17 amino acid peptide near the center of human PRKCDBP. The immunogen is located within amino acids 150 - 200 of PRKCDBP. Amino Acid Squence: ESSDEEPVESRAQRLRRT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 112464
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.