missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKCBP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37981-20ul
This item is not returnable.
View return policy
Description
PRKCBP1 Polyclonal antibody specifically detects PRKCBP1 in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| PRKCBP1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| ESP-2, HED-2, zyxin, Zyxin-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 959-1188 of human PRKCBP1 (NP_898868.1).,, Sequence:, LELTMAEMRQSLEQERDRLIAEVKKQLELEKQQAVDETKKKQWCANCKKEAIFYCCWNTSYCDYPCQQAHWPEHMKSCTQSATAPQQEADAEVNTETLNKSSQGSSSSTQSAPSETASASKEKETSAEKSKESGSTLDLSGSRETPSSILLGSNQGSVSKRCDKQPAYAPTTTDHQPHPNYPAQKYHSRSNKSSWSSSDEKRGSTRSDHNTSTSTKSLLPKESRLDTFWD | |
| 20 μL | |
| Signal Transduction | |
| 23613 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction