missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKCBP1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-19131AF594
This item is not returnable.
View return policy
Description
PRKCBP1 Polyclonal antibody specifically detects PRKCBP1 in Human samples. It is validated for Western Blot
Specifications
| PRKCBP1 | |
| Polyclonal | |
| Western Blot | |
| ESP-2, HED-2, zyxin, Zyxin-2 | |
| The immunogen recognized by this antibody maps to a region between residue 1136 and 1186 of human protein kinase C binding protein 1 (zinc finger, MYND-type containing 8) using the numbering given in entry CAI21842.1 (GeneID 23613). | |
| 0.1 mL | |
| Signal Transduction | |
| 23613 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction