missing translation for 'onlineSavingsMsg'
Learn More

PRKAR2B Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech

Product Code. 17291396 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17291396 20 μL 20µL
17281396 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17291396 Supplier Proteintech Supplier No. 283511AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The PRKAR2B antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human PRKAR2B. This antibody recognizes human, mouse, rat antigen. The PRKAR2B antibody has been validated for the following applications: WB,ELISA analysis.

Specifications

Antigen PRKAR2B
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 650 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 5577
Target Species Human, Mouse, Rat
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.