missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKAR2B, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB31668.100uL
This item is not returnable.
View return policy
Specifications
| PRKAR2B | |
| Polyclonal | |
| ELISA (1:10000) Immunohistochemistry (1:100-300) Western Blot (1:500-2000) The optimal working dilution should be determined by the end user. | |
| protein kinase, cAMP-dependent, regulatory, type II, beta | |
| PRKAR2B | |
| A synthetic peptide corresponding to amino acids 50-130 of human PRKAR2B (phospho S113). | |
| RUO | |
| 5577 | |
| Human, Monkey, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| In PBS (50% glycerol, 0.5% BSA and 0.02% sodium azide). | |
| PRKAR2/RII-BETA | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody detects endogenous levels of PKAII beta reg protein only when phosphorylated at S113. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction