missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRKAR2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106470
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PKA R2.
cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase (AMPK), which transduces the signal through phosphorylation of different target proteins. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans. The protein encoded by this gene is one of the regulatory subunits. This subunit can be phosphorylated by the activated catalytic subunit. It may interact with various A-kinase anchoring proteins and determine the subcellular localization of AMPK.
Specifications
| PRKAR2A | |
| Polyclonal | |
| Unconjugated | |
| PRKAR2A | |
| 1110061A24Rik; AI317181; AI836829; cAMP-dependent protein kinase regulatory subunit RII alpha; cAMP-dependent protein kinase type II-alpha regulatory subunit; cAMP-dependent protein kinase, regulatory subunit alpha 2; MGC3606; PKR2; PRKAR2; Prkar2a; protein kinase A, RII-alpha subunit; protein kinase cAMP-dependent regulatory type II alpha; protein kinase cAMP-dependent type II regulatory subunit alpha; protein kinase, cAMP dependent regulatory, type II alpha; protein kinase, cAMP-dependent, regulatory subunit type II alpha; protein kinase, cAMP-dependent, regulatory, type 2, alpha; protein kinase, cAMP-dependent, regulatory, type II, alpha; RII(alpha); unnamed protein product | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19087, 29699, 5576 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P12367, P12368, P13861 | |
| PRKAR2A | |
| A synthesized peptide derived from human PRKAR2A(Accession P13861), corresponding to amino acid residues S48-V98. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction