missing translation for 'onlineSavingsMsg'
Learn More

PRKAG3 Antibody, Novus Biologicals™

Product Code. 18325579 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325579 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325579 Supplier Novus Biologicals Supplier No. H00053632D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRKAG3 Polyclonal antibody specifically detects PRKAG3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRKAG3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.4)
Gene Accession No. NP_059127.2
Gene Alias 5'-AMP-activated protein kinase subunit gamma-3, AMPK gamma3, AMPK gamma-3 chain, AMPK subunit gamma-3, AMPKG3,5'-AMP-activated protein kinase, gamma-3 subunit, protein kinase, AMP-activated, gamma 3 non-catalytic subunit
Host Species Rabbit
Immunogen PRKAG3 (NP_059127.2, 1 a.a. - 489 a.a.) full-length human protein. MEPGLEHALRRTPSWSSLGGSEHQEMSFLEQENSSSWPSPAVTSSSERIRGKRRAKALRWTRQKSVEEGEPPGQGEGPRSRPAAESTGLEATFPKTTPLAQADPAGVGTPPTGWDCLPSDCTASAAGSSTDDVELATEFPATEAWECELEGLLEERPALCLSPQAPFPKLGWDDELRKPGAQIYMRFMQEHTCYDAMATSSKLVIFDTMLEIKKAFFALVANGVRAAPLWDSKKQSFVGMLTITDFILVLHRYYRSPLVQIYEIEQHKIETWREIYLQGCFKPLVSISPNDSLFEAVYTLIKNRIHRLPVLDPVSGNVLHILTHKRLLKFLHIFGSLLPRPSFLYRTIQDLGIGTFRDLAVVLETAPILTALDIFVDRRVSALPVVNECGQVVGLYSRFDVIHLAAQQTYNHLDMSVGEALRQRTLCLEGVLSCQPHESLGEVIDRIAREQVHRLVLVDETQHLLGVVSLSDILQALVLSPAGIDALGA
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 53632
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.