missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKAG3 Antibody (2C7A7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61796-0.025ml
This item is not returnable.
View return policy
Description
PRKAG3 Monoclonal specifically detects PRKAG3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PRKAG3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| 5'-AMP-activated protein kinase subunit gamma-3, AMPK gamma3, AMPK gamma-3 chain, AMPK subunit gamma-3, AMPKG3,5'-AMP-activated protein kinase, gamma-3 subunit, protein kinase, AMP-activated, gamma 3 non-catalytic subunit | |
| Mouse | |
| 54.3 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 2C7A7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PRKAG3 | |
| Purified recombinant fragment of human PRKAG3 (AA: 9-151) expressed in E. Coli. | |
| Protein G purified | |
| Signal Transduction | |
| 53632 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction