missing translation for 'onlineSavingsMsg'
Learn More

PRKACG Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55346-1-AP-150UL

 View more versions of this product

Product Code. 16846825

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cyclic AMP-dependent protein kinase (PKA) consists of two catalytic subunits and a regulatory subunit dimer. This gene encodes the gamma form of its catalytic subunit. The gene is intronless and is thought to be a retrotransposon derived from the gene for the alpha form of the PKA catalytic subunit.
Specifications

Specifications

PRKACG
Polyclonal
Unconjugated
PRKACG
KAPG, PKA C gamma, PKACg, PRKACG
Rabbit
Antigen Affinity Chromatography
RUO
5568
-20°C
Liquid
Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P22612
PRKACG
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.