missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKACG Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55346-1-AP-20UL
This item is not returnable.
View return policy
Description
Cyclic AMP-dependent protein kinase (PKA) consists of two catalytic subunits and a regulatory subunit dimer. This gene encodes the gamma form of its catalytic subunit. The gene is intronless and is thought to be a retrotransposon derived from the gene for the alpha form of the PKA catalytic subunit.Specifications
| PRKACG | |
| Polyclonal | |
| Unconjugated | |
| PRKACG | |
| KAPG, PKA C gamma, PKACg, PRKACG | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5568 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P22612 | |
| PRKACG | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction