Learn More
Pritoxaximab Recombinant Human Monoclonal Antibody
GREENER_CHOICE
Pritoxaximab Recombinant Monoclonal Antibody
Brand:  Invitrogen MA560165
 View more versions of this product
Product Code. 30284502
  • £397.00 / 100µg
Description
Pritoxaximab Recombinant Monoclonal Antibody for SPR, Flow, ELISA, Functional
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pritoxaximab
Recombinant Monoclonal
Unconjugated
caStx1; Shigamabs
Protein A
RUO
Bacteria
Antibody
IgG1
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized