missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pristinamycin IA, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
Description
Pristinamycin IA (Mikamycin B) is a cycle-peptidic macrolactone antibiotic. Pristinamycin IA is a substrate of P-glycoprotein and inhibits its function. Pristinamycin IA is active against StaphyloEoccus and Srreptococcus.
Specifications
Specifications
| Chemical Name or Material | Pristinamycin IA |
| Physical Form | Powder |
| CAS | 3131-03-1 |
| Color | Yellow |
| Quantity | 25 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C45H54N8O10 |
| Synonym | Mikamycin B Mikamycin IA |
| Solubility Information | DMSO : 33.33 mg/mL (38.44 mM; Need ultrasonic) |
| SMILES | O=C(N[C@@H]1C(N[C@H](CC)C(N2[C@](CCC2)([H])C(N(C)[C@@H](CC3=CC=C(N(C)C)C=C3)C(N4[C@](CC(CC4)=O)([H])C(N[C@@H](C5=CC=CC=C5)C(O[C@@H]1C)=O)=O)=O)=O)=O)=O)C6=NC=CC=C6O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction