missing translation for 'onlineSavingsMsg'
Learn More

Pristinamycin IA, MedChemExpress

Product Code. 16462029
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16462029 1 mg 1mg
16458318 10 mg 10mg
16433786 25 mg 25mg
16420758 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16462029 Supplier Medchem Express Supplier No. HYA0279A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Pristinamycin IA (Mikamycin B) is a cycle-peptidic macrolactone antibiotic. Pristinamycin IA is a substrate of P-glycoprotein and inhibits its function. Pristinamycin IA is active against StaphyloEoccus and Srreptococcus.

Pristinamycin IA (Mikamycin B) is a cycle-peptidic macrolactone antibiotic. Pristinamycin IA is a substrate of P-glycoprotein and inhibits its function. Pristinamycin IA is active against StaphyloEoccus and Srreptococcus.

Specifications

Chemical Name or Material Pristinamycin IA
Physical Form Powder
CAS 3131-03-1
Color Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C45H54N8O10
Synonym Mikamycin B Mikamycin IA
Solubility Information DMSO : 33.33 mg/mL (38.44 mM; Need ultrasonic)
SMILES O=C(N[C@@H]1C(N[C@H](CC)C(N2[C@](CCC2)([H])C(N(C)[C@@H](CC3=CC=C(N(C)C)C=C3)C(N4[C@](CC(CC4)=O)([H])C(N[C@@H](C5=CC=CC=C5)C(O[C@@H]1C)=O)=O)=O)=O)=O)=O)C6=NC=CC=C6O
Molecular Weight (g/mol) 866.96
Formula Weight 866.96
Percent Purity 96.32%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.