missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prion protein, Mouse anti-Human,Hamster, Clone: 3F4, Novus Biologicals™
Mouse Monoclonal Antibody
£217.00 - £489.00
Specifications
| Antigen | Prion protein |
|---|---|
| Clone | 3F4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18258128
|
Novus Biologicals
NBP2-81078-0.025MG |
0.025 mg |
£217.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18054006
|
Novus Biologicals
NBP2-81078-0.2MG |
0.2 mg |
£489.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PrPSc Monoclonal specifically detects PrPSc in Human, Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Prion protein | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Neuroscience, Stem Cell Markers | |
| PBS | |
| 5621 | |
| Hamster 263K PrPs. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 3F4 | |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Hamster | |
| CD230, CJD, fatal familial insomnia), GSS, prion protein, prion protein (p27-30), prion protein PrP, prion-related protein, PRIPMGC26679 | |
| PRNP | |
| IgG1 κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title