Learn More
Epredia™ PrintMate™ Embedding Cassettes in Tubes, Aqua
Contain incised tissue safely and securely for tissue processing, embedding and sectioning with Epredia™ PrintMate™ Cassettes.
Brand: Epredia™ A84210011
Code : NEW
Additional Details : Weight : 4.50000kg
Description
PrintMate Embedding Cassettes in pre-packed tubes, Blue. 20 tubes X 75 cassettes, total 1500 cassettes.Specifications
Round Pore | |
Yes | |
Embedding Biopsy Cassette |
20 Tubes X 75 Cassettes, total 1500 Cassettes. | |
Blue |