Learn More
Epredia™ PrintMate™ Embedding Cassettes with Lids, aqua
Product Code. 15487624 Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Product Code. 15487624
Brand: Epredia™ A84210070
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
TRUSTED_SUSTAINABILITY
Specifications
PrintMate Compatible Yes
Color Aqua