missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ PrintMate™ Embedding Cassettes

Product Code. 16104027 Shop All Epredia Products
Change view
Click to view available options
Color:
Aqua
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Quantity:
10 x 75 cassettes (750)/cs
1000
4 x 250 cassettes (1000)/cs
Closure Included:
No lids
Separate lids included
Unit Size:
Pack of 1000
Pack of 750
57 product options available for selection
Product selection table with 57 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity Closure Included unitSize
16104027 Yellow 10 x 75 cassettes (750)/cs No lids Pack of 750
15487594 Aqua - - Pack of 1000
16363946 Beige 10 x 75 cassettes (750)/cs No lids Pack of 750
15497594 Blue - - Pack of 1000
16373946 Blue 10 x 75 cassettes (750)/cs No lids Pack of 750
15407604 Green - - Pack of 1000
16383946 Green 10 x 75 cassettes (750)/cs No lids Pack of 750
15417604 Gray - - Pack of 1000
16393946 Gray 10 x 75 cassettes (750)/cs No lids Pack of 750
15427604 Lilac - - Pack of 1000
16303956 Lavender 10 x 75 cassettes (750)/cs No lids Pack of 750
15437604 Orange - - Pack of 1000
16313956 Light Green 10 x 75 cassettes (750)/cs No lids Pack of 750
15447604 Pink - - Pack of 1000
16323956 Lilac 10 x 75 cassettes (750)/cs No lids Pack of 750
15457604 Tan - - Pack of 1000
16333956 Orange 10 x 75 cassettes (750)/cs No lids Pack of 750
15471820 White - - Pack of 1000
16343956 Pink 10 x 75 cassettes (750)/cs No lids Pack of 750
15477604 Yellow 1000 - Pack of 1000
16353956 Tan 10 x 75 cassettes (750)/cs No lids Pack of 750
16363956 White 10 x 75 cassettes (750)/cs No lids Pack of 750
16373956 Beige 4 x 250 cassettes (1000)/cs No lids Pack of 1000
16383956 Blue 4 x 250 cassettes (1000)/cs No lids Pack of 1000
16393956 Green 4 x 250 cassettes (1000)/cs No lids Pack of 1000
16303966 Gray 4 x 250 cassettes (1000)/cs No lids Pack of 1000
16313966 Lavender 4 x 250 cassettes (1000)/cs No lids Pack of 1000
16323966 Light Green 4 x 250 cassettes (1000)/cs No lids Pack of 1000
16333966 Lilac 4 x 250 cassettes (1000)/cs No lids Pack of 1000
16343966 Orange 4 x 250 cassettes (1000)/cs No lids Pack of 1000
16353966 Pink 4 x 250 cassettes (1000)/cs No lids Pack of 1000
16363966 Tan 4 x 250 cassettes (1000)/cs No lids Pack of 1000
16373966 White 4 x 250 cassettes (1000)/cs No lids Pack of 1000
16383966 Yellow 4 x 250 cassettes (1000)/cs No lids Pack of 1000
16393966 Beige 4 x 250 cassettes (1000)/cs Separate lids included Pack of 1000
16303976 Blue 4 x 250 cassettes (1000)/cs Separate lids included Pack of 1000
16313976 Green 4 x 250 cassettes (1000)/cs Separate lids included Pack of 1000
16323976 Gray 4 x 250 cassettes (1000)/cs Separate lids included Pack of 1000
16333976 Lavender 4 x 250 cassettes (1000)/cs Separate lids included Pack of 1000
16343976 Light Green 4 x 250 cassettes (1000)/cs Separate lids included Pack of 1000
16353976 Lilac 4 x 250 cassettes (1000)/cs Separate lids included Pack of 1000
16363976 Orange 4 x 250 cassettes (1000)/cs Separate lids included Pack of 1000
16373976 Pink 4 x 250 cassettes (1000)/cs Separate lids included Pack of 1000
16383976 Tan 4 x 250 cassettes (1000)/cs Separate lids included Pack of 1000
16393976 White 4 x 250 cassettes (1000)/cs Separate lids included Pack of 1000
16303986 Yellow 4 x 250 cassettes (1000)/cs Separate lids included Pack of 1000
16344006 Beige 10 x 75 cassettes (750)/cs Separate lids included Pack of 750
16354006 Blue 10 x 75 cassettes (750)/cs Separate lids included Pack of 750
16364006 Green 10 x 75 cassettes (750)/cs Separate lids included Pack of 750
16374006 Gray 10 x 75 cassettes (750)/cs Separate lids included Pack of 750
16384006 Lavender 10 x 75 cassettes (750)/cs Separate lids included Pack of 750
16394006 Light Green 10 x 75 cassettes (750)/cs Separate lids included Pack of 750
16304016 Lilac 10 x 75 cassettes (750)/cs Separate lids included Pack of 750
16314016 Orange 10 x 75 cassettes (750)/cs Separate lids included Pack of 750
16324016 Pink 10 x 75 cassettes (750)/cs Separate lids included Pack of 750
16334016 Tan 10 x 75 cassettes (750)/cs Separate lids included Pack of 750
16344016 White 10 x 75 cassettes (750)/cs Separate lids included Pack of 750
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 57 options available.
57 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16104027 missing translation for 'mfr' Epredia™ missing translation for 'supplierNo' B851010YW

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.

Designed for performance and user convenience, PrintMate Embedding Cassettes have been optimized for superior workflow and printing with the PrintMate Classic and PrintMate AS printing systems. The even, textured printing face provides the optimal surface for high-quality printing every time. For additional information, the right and left sides are textured for clear handwriting with suitable pens. The lid catches have been adjusted for ease of use with low opening and closing force, while still giving the security needed while processing.Available in tubes for automatic printing or loose for manual use, and in twelve colors for enhanced workflow identification.PrintMate Embedding Cassettes are also available without lids, for economical use with reuseable metal lids.
TRUSTED_SUSTAINABILITY

Specifications

Pore Style Round Holes
Closure Included No lids
Quantity 10 x 75 cassettes (750)/cs
Closure Type Reversible
PrintMate Compatible Yes
Color Yellow
For Use With (Equipment) For Use with Metal Lids
Product Type Embedding Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.