Learn More
Epredia™ PrintMate™ Biopsy Cassettes in Tubes, Aqua
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. Packed in tubes, PrintMate Cassettes are intended for high throughput printing, and provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Brand: Epredia™ A84210024
Code : NEW
Additional Details : Weight : 4.50000kg
Description
PrintMate Biopsy Cassettes in pre-packed tubes. 20 tubes X 75 cassettes, total 1500 cassettes.Specifications
Small Square Pore | |
Yes | |
Lilac | |
Biopsy Cassette |