Description
Prints each cassette in 10 seconds or less
Small footprint allows the printer to fit easily next to the grossing station
Print only the cassettes you need, right at the point of use Print barcodes on cassettes to automate downstream processes Eliminate manual handwriting processes
Small footprint allows the printer to fit easily next to the grossing station
Print only the cassettes you need, right at the point of use Print barcodes on cassettes to automate downstream processes Eliminate manual handwriting processes
Specifications
Specifications
| Description | PrintMate AS 450 Cassette Printer |
| Capacity | 450 Cassettes |
| Includes | PrintMate AS USB cable Power cable 6 cassette hoppers One roll of tape (installed) Epredia™ LabWriter™ software CD Operator guide |
| Electrical Requirements | 100/240 VAC, 50/60 Hz, 150 VA (512 BTU/h) |
| Product Type | Cassette Printer |