Learn More
Hamilton™ Priming Kit
Description
Specification Sheet
- Consists of – one 250 uL syringe (p/n 81120), one 6 pack of 30 gauge needles, and one pack of septa (p/n 75826).
- Compatible Syringes – The priming kit is compatible with 600, 700, 1700, removable needle syringes and 7000 series syringes
Specifications
Specifications
| For Use With (Equipment) | The Priming Kit is Compatible with 600, 700, 1700, Removable Needle Syringes and 7000 Series Syringes |