missing translation for 'onlineSavingsMsg'
Learn More

Primidone, MedChemExpress

Product Code. 16458246
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16458246

Brand: Medchem Express HYB0339

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Primidone is a potent anticonvulsant agent of the barbiturate class. Primidone is a neuronal voltage-gated sodium channel (VGSC) blocker and can be used for the study of epilepsy, essential tremor, and Psychiatric disorders.

Primidone is a potent anticonvulsant agent of the barbiturate class. Primidone is a neuronal voltage-gated sodium channel (VGSC) blocker and can be used for the study of epilepsy, essential tremor, and Psychiatric disorders.

Specifications

Chemical Name or Material Primidone
Physical Form Solid
CAS 125-33-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H14N2O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 50 mg/mL (229.10 mM; Need ultrasonic) ∣H2O : ≥ 0.67 mg/mL (3.07 mM)
SMILES O=C(C1(CC)C2=CC=CC=C2)NCNC1=O
Molecular Weight (g/mol) 218.25
Formula Weight 218.25
Percent Purity 99.82%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.