missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Primidone, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Primidone |
| CAS | 125-33-7 |
| Analyte or Component Names | Primidone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H14 N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Primidone, 5-Ethyl-5-phenyldihydropyrimidine-4,6(1H,5H)-dione, 4,6(1H,5H)-Pyrimidinedione, 5-ethyldihydro-5-phenyl- (6CI,8CI,9CI), 2-Deoxyphenobarbital, 2-Desoxyphenobarbital, 5-Ethyl-5-phenylhexahydropyrimidine-4,6-dione, 5-Ethyldihydro-5-phenyl-4,6(1H,5H)-pyrimidinedione, 5-Ethylhexahydro-5-phenylpyrimidine-4,6-dione, 5-Phenyl-5-ethyl-hexahydropyrimidine-4,6-dione, Hexamidine, Hexamidine (antiepileptic), Hexamydin, Lepimidin, Liskantin, Milepsin, Misodine, Misolyne, Mizodin, Mizolin, Mylepsin, Mysedon, Mysoline, NSC 41701, Neurosyn, Primaclone, Primacone, Primakton, Primidon, Pyrimidone Medi-pets, Resimatil, Roe 101, Sertan |
| InChI Formula | InChI=1S/C12H14N2O2/c1-2-12(9-6-4-3-5-7-9)10(15)13-8-14-11(12)16/h3-7H,2,8H2,1H3,(H,13,15)(H,14,16) |
| Solution Type | Neat |
| SMILES | CCC1(C(=O)NCNC1=O)c2ccccc2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction