missing translation for 'onlineSavingsMsg'
Learn More

Primidone EP Impurity C, Mikromol™

Product Code. 30050234
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30050234

Brand: Mikromol MM1030.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Primidone EP Impurity C
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-2-Phenylbutanamide
CAS 90-26-6
Analyte or Component Names (2RS)-2-Phenylbutanamide
Shipping Condition Room Temperature
Molecular Formula C10 H13 N O
Certifications/Compliance ISO 9001
Synonym Lactam of S-Carboxymethyl-L-cysteinamide, Primidone Imp. C (EP), Primidone USP Related Compound C, Primidone USP RC C, (2RS)-2-Phenylbutanamide, Primidone Related Compound C, Primidone Impurity C
InChI Formula InChI=1S/C10H13NO/c1-2-9(10(11)12)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3,(H2,11,12)
SMILES CCC(C(=O)N)c1ccccc1
IUPAC Name 2-phenylbutanamide
Molecular Weight (g/mol) 163.22
Quantity 100 mg
Formula Weight 163.0997 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.