missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PrimeStore Molecular Transport Medium

Product Code. 17808377
Click to view available options
Fill Volume:
1.5 mL
Tube Size:
5 mL
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 17808377

Brand: Thermo Scientific™ R13905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Non-hazardous, inactivation media for viral collection and transport with Inactivation media for nucleic acid testing and molecular diagnostics.

Stabilize, inactivate and transport infectious unprocessed nasal washes, nasopharyngeal, oral / throat swabs suspected of containing influenza A virus RNA with Longhorn PrimeStore Molecular Transport Medium™ (PS-MTM).

PrimeStore Molecular Transport Medium™ is also intended for the stabilization, transportation, and inactivation of infectious unprocessed sputum samples suspected of containing Mycobacterium tuberculosis (MTB) DNA from human samples.

Specimen collection and transport is a key component in the molecular detection of influenza A and MTB. PS-MTM is a self-contained, ‘ready-to-use’ system that allows for the stabilization and safe transport of clinical samples at ambient temperature from the collection site to the laboratory.

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Compatibility The PS-MTM system is a collection, preservation, transport, and storage system for influenza RNA and MTB DNA. Extraction and purification of nucleic acids have been validated on several manual spin column kits (PrimeXtract™, RNAqueous Micro Kit, Viral RNA Mini Kit, QiaAMP DNA Mini Kit) and automated magnetic bead extraction kits (NucliSENS EasyMAG and MagNA Pure 96 System using the DNA Bacterial/Viral small volume kit). The user is responsible for validating additional extraction and purification kits and platforms.
Description PrimeStore MTM Collection Tube (1.5mL), 50/bx
Fill Volume 1.5 mL
For Use With (Application) Intended for stabilization, transportation, and inactivation of infectious unprocessed nasal washes, nasopharyngeal, oral/throat swabs suspected of containing influenza A virus RNA. PS-MTM is also intended for the stabilization, transportation and inactivation of infectious unprocessed sputum samples suspected of containing Mycobacterium tuberculosis (MTB) DNA from human samples.
Quantity 50/Pk.
Storage Requirements Room temperature, 2°C to 25°C
Tube Size 5 mL

Distributed globally by Thermo Fisher Scientific

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.