Learn More
Invitrogen™ PRIMER, 5'ALEXA 680
Product Code. 10262999
Click to view available options
:
50 nmol
For Research Use Only. All usage must comply with product instructions.
Product Code. 10262999
Brand: Invitrogen™ 100007337
For Research Use Only. All usage must comply with product instructions.
Custom Primer
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Description 5' Alexa 680 Primer, 50 nmol
Product Type 5' Alexa 680 Primer
Primer 5' Labeled
Quantity 50 nmol