missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PrimeFlow™ Fixation/Permeabilization Buffer Set
Description
Includes
- PrimeFlow™ RNA Fixation Buffer 1A: cat. 00-18100
- PrimeFlow™ RNA Fixation Buffer 1B: cat. 00-18200
- PrimeFlow™ RNA Permeabilization Buffer (10X): cat. 00-18300
- PrimeFlow™ RNA Fixation Buffer 2 (8X): cat. 00-18400
- PrimeFlow™ RNA Wash Buffer: cat. 00-19180
Note that additional changes to antibody performance may be observed after the hybridization steps of the PrimeFlow™ RNA assay.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Intracellular Staining Followed by Flow Cytometric Analysis |
| For Use With (Equipment) | Flow cytometers |
| No. of Reactions | 100 Tests |
| Product Type | Buffer Set for RNA Assay |
| Quantity | 100 Tests |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?