missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Monkey P-Selectin/CD62P ELISA Kit (Colorimetric)

Product Code. 18084447 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18084447 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18084447 Supplier Novus Biologicals™ Supplier No. NBP192719

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Primate P-Selectin/CD62P ELISA Kit (Colorimetric) measures primate P-Selectin/CD62P in cell culture supernatant, serum (baboon).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.8 - 50 U/mL
Assay Sensitivity 1.3 U/mL
Product Type ELISA Kit
Sample Type Baboon Serum, Cell Culture Supernatant
Specificity The interference of circulating factors of the immune systemwas evaluated by spiking these proteins at physiologically relevant concentrations into a monkey P-Selectin/CD62P positive serum. There was no crossreactivity detected.
Target Species Primate
For Use With (Application) ELISA
Gene ID (Entrez) 6403
Gene Symbol SELP
Kit Contents Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey sP-selectin, HRP-Conjugate anti-monkey sP-selectin monoclonal antibody, Monkey sP-selectin Standard lyophilized, 100.0U/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
Quantity 96 Tests
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CD62P antigen, FLJ45155, PADGEMantigen CD62), selectin P (granule membrane protein 140kDa, antigen CD62)
Target P-Selectin/CD62P
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.