missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Monkey ICAM-1/CD54 ELISA Kit (Colorimetric)

Product Code. 18028516 Shop All Bio Techne Products
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 18028516

Brand: Novus Biologicals™ NBP192712

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 

The Human, Primate ICAM-1/CD54 ELISA Kit (Colorimetric) measures human, primate ICAM-1/CD54 in cell culture supernatant, serum (baboon, rhesus serum).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 6.3 - 100 U/mL
Assay Sensitivity 1.1 U/mL
Product Type ELISA Kit
Sample Type Baboon Serum, Cell Culture Supernatant, Rhesus Serum
Target Species Primate
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 3383
Gene Symbol ICAM1
Kit Contents HRP-Conjugate anti-monkey sICAM-1 monoclonal antibody, Monkey sICAM-1 Standard, 100U/mL, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films, Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey sICAM-1
Quantity 96 Tests
Research Discipline Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Target ICAM-1/CD54
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.