missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Primary Hepatocyte Maintenance Supplements

Product Code. 10263580
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10263580

Brand: Gibco™ CM4000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Primary Hepatocyte Maintenance Supplements

Hepatocyte Maintenance Supplement Pack

Hepatocyte Maintenance Supplement Packs allow you to supplement up to 500 ml of Williams Medium E without phenol red (or suitable alternative basal medium) for the purpose of incubating hepatocytes in suspension or plated cultures.

Hepatocyte Maintenance Supplement Packs contain:

  • Dexamethasone
  • A cocktail solution of penicillin-streptomycin, ITS+ (insulin, transferrin, selenium complex, BSA, and linoleic acid), GlutaMAX™, and HEPES

    For research use only. Not intended for human or animal therapeutic or diagnostic use.

    Related Links:

    Learn more about our cell culture reagents for hepatocytes.

    Learn more about our cryopreserved hepatocytes.

    Learn about our full line of hepatocytes.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store Cocktail B and dexamethasone in refrigerator (2–8°C) and protect from light.
    Cell Type Hepatocyte Cells
    Culture Type Mammalian Cell Culture, Insect Cell Culture
    Form Cryopreserved
    Product Type Hepatocyte Supplement
    With Additives GlutaMAX, HEPES
    Without Additives No Phenol Red
    Product Line Gibco
    Quantity 1 kit
    compliance-icons
    Product Identifier
    • Primary Hepatocyte Maintenance Supplements
    Signal Word
    • Warning
    Hazard Category
    • Reproductive toxicity Category 2
    Hazard Statement
    • H361-Suspected of damaging fertility or the unborn child.
    Precautionary Statement
    • P201-Obtain special instructions before use.
    • P202-Do not handle until all safety precautions have been read and understood.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P308+P313-IF exposed or concerned: Get medical advice/attention.
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
    Supplemental information
    • MIXTURE LIST-Contains : streptomycin sulphate

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.