missing translation for 'onlineSavingsMsg'
Learn More

Primaquine diphosphate, 98%

Product Code. 10756832 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10756832 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10756832 Supplier Thermo Scientific Acros Supplier No. 170870010
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63-45-6 | C15H21N3O·2H3PO4 | 455.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 63-45-6
Melting Point 205.0°C to 206.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C15H21N3O·2H3PO4
MDL Number MFCD00013166
Quantity 1 g
Merck Index 15,7863
Synonym primaquine phosphate, n4-6-methoxyquinolin-8-yl pentane-1,4-diamine phosphate, n-5-aminopentan-2-yl-6-methoxyquinolin-8-amine; phosphoric acid, 4-n-6-methoxyquinolin-8-yl pentane-1,4-diamine; phosphoric acid, primaquine diphosphate, chembl994, phosphoric acid; primaquine, 1, n4-6-methoxy-8-quinolinyl-, phosphate 1:2, n-5-aminopentan-2-yl-6-methoxyquinolin-8-amine phosphoric acid, n4-6-methoxyquinolin-8-yl pentane-1,4-diamine; phosphoric acid
Solubility Information Solubility in water: moderately soluble.
InChI Key GJOHLWZHWQUKAU-UHFFFAOYSA-N
SMILES CC(CCCN)NC1=C2C(=CC(=C1)OC)C=CC=N2.OP(=O)(O)O.OP(=O)(O)O
IUPAC Name 4-N-(6-methoxyquinolin-8-yl)pentane-1,4-diamine;phosphoric acid
Molecular Weight (g/mol) 455.34
PubChem CID 6135
Formula Weight 455.34
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Primaquine diphosphate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.