missing translation for 'onlineSavingsMsg'
Learn More

Primaquine, 98.81%, MP Biomedicals™

Product Code. 11463742 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463742 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11463742 Supplier MP Biomedicals Supplier No. 0215636610
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 63-45-6
Molecular Formula C15H27N3O9P2
Quantity 10 g
Synonym 8-[4-Amino-1-methylbutylamino]-6-methoxyquinoline
InChI Key GJOHLWZHWQUKAU-UHFFFAOYNA-N
SMILES OP(O)(O)=O.OP(O)(O)=O.COC1=CC(NC(C)CCCN)=C2N=CC=CC2=C1
IUPAC Name N4-(6-methoxyquinolin-8-yl)pentane-1,4-diamine; bis(phosphoric acid)
Molecular Weight (g/mol) 455.34
Formula Weight 455.3
Physical Form Crystalline Powder
Chemical Name or Material Primaquine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.