missing translation for 'onlineSavingsMsg'
Learn More

PRIMA-1, Tocris Bioscience™

Product Code. 18085132
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18085132

Brand: Tocris Bioscience 1862/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Restores mutant p53 activity; induces apoptosis

Selectively restores mutant p53 activity in tumor cells via activation of Bax and PUMA. Induces apoptosis and inhibits growth of tumors with mutant p53.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5608-24-2
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
InChI Key RFBVBRVVOPAAFS-UHFFFAOYSA-N
Synonym prima-1, 2,2-bis hydroxymethyl-1-azabicyclo 2,2,2, octan-3-one, 2,2-bis hydroxymethyl-1-azabicyclo 2.2.2 octan-3-one, 2,2-bis hydroxymethyl-3-quinuclidinone, p53 reactivation and induction of massive apoptosis, 2,2-bishydroxymethyl-1-aza-bicyclo 2.2.2 octan-3-one, 2,2-bis hydroxymethyl-1-azabicyclo 2,2,2 octan-3-one, prima 1, tocris-1862, prima 1;prima1
PubChem CID 322968
IUPAC Name 2,2-bis(hydroxymethyl)-1-azabicyclo[2.2.2]octan-3-one
SMILES C1CN2CCC1C(=O)C2(CO)CO

Specifications

Quantity 10 mg
Formula Weight 185.22
Percent Purity >99%
Chemical Name or Material PRIMA-1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.