missing translation for 'onlineSavingsMsg'
Learn More

Prima-1, Thermo Scientific Chemicals

Product Code. 15476179
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476179 10 mg 10mg
15486179 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476179 Supplier Thermo Scientific Alfa Aesar Supplier No. J64071.MA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cell-permeable quinuclidinone analog

A cell-permeable quinuclidinone analog.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5608-24-2
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD04974196
InChI Key RFBVBRVVOPAAFS-UHFFFAOYSA-N
Synonym prima-1, 2,2-bis hydroxymethyl-1-azabicyclo 2,2,2, octan-3-one, 2,2-bis hydroxymethyl-1-azabicyclo 2.2.2 octan-3-one, 2,2-bis hydroxymethyl-3-quinuclidinone, p53 reactivation and induction of massive apoptosis, 2,2-bishydroxymethyl-1-aza-bicyclo 2.2.2 octan-3-one, 2,2-bis hydroxymethyl-1-azabicyclo 2,2,2 octan-3-one, prima 1, tocris-1862, prima 1;prima1
PubChem CID 322968
IUPAC Name 2,2-bis(hydroxymethyl)-1-azabicyclo[2.2.2]octan-3-one
SMILES C1CN2CCC1C(=O)C2(CO)CO

Specifications

Color White
Quantity 10 mg
Sensitivity Light sensitive
Solubility Information Soluble in water at 10mg/ml
Formula Weight 185.2
Physical Form Solid
Chemical Name or Material Prima-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.